Anti-KRT18 Mouse Monoclonal Antibody (CF488A) [clone: KRT18/835]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC880835-500 BNC880835-100
BNC880835-500EA 560 EUR
BNC880835-500 BNC880835-100
Anti-KRT18 Mouse Monoclonal Antibody (CF488A) [clone: KRT18/835]
Antilichamen
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It is reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18.
Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: KRT18/835
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human