Anti-CDC2 Mouse Monoclonal Antibody (CF568) [clone: POH-1]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC680853-500 BNC680853-100
BNC680853-500EA 560 EUR
BNC680853-500 BNC680853-100
Anti-CDC2 Mouse Monoclonal Antibody (CF568) [clone: POH-1]
Antilichamen
Recognizes a 34 kDa protein, which is identified as cyclin dependent kinase 1 (cdk1) or p34cdc2 protein kinase. cdk1 / p34cdc2 plays a crucial role during cell division and is most active during mitosis. It is predominantly localized in the nucleus. It is a serine/threonine kinase, which is activated by cyclin, presumably by de-phosphorylation of tyrosine residues. Activated cdk1 / p34cdc2 performs specific functions during mitosis, including nuclear envelope breakdown and chromosome condensation.

Type: Primary
Antigen: CDC2
Clonality: Monoclonal
Clone: POH-1
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Human, Mink, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR