Anti-RBP1 Mouse Monoclonal Antibody (Purified) [clone: RBP/872]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNUB0872-500 BNUB0872-100
BNUB0872-500EA 511 EUR
BNUB0872-500 BNUB0872-100
Anti-RBP1 Mouse Monoclonal Antibody (Purified) [clone: RBP/872]
Antilichamen
Recognizes a protein of 21 kDa-25 kDa, identified as retinol binding protein-1 (RBP1). This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post-transnationally and results in defective delivery and supply to the epidermal cells.
Type: Primary
Antigen: RBP1
Clonality: Monoclonal
Clone: RBP/872
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Goat, Human, Monkey, Mouse, Rabbit, Rat