Anti-RBP1 Mouse Monoclonal Antibody (Biotin) [clone: RBP/872]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNCB0872-500 BNCB0872-100
BNCB0872-500EA 560 EUR
BNCB0872-500 BNCB0872-100
Anti-RBP1 Mouse Monoclonal Antibody (Biotin) [clone: RBP/872]
Antilichamen
Recognizes a protein of 21 kDa-25 kDa, identified as retinol binding protein-1 (RBP1). This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post-transnationally and results in defective delivery and supply to the epidermal cells.

Type: Primary
Antigen: RBP1
More Product Information
Order Now

Specificaties

antigeen symbool RBP1
Type antilichaam Primair,Retinol binding protein 1
Environmentally Preferable
Type antilichaam Primair,Retinol binding protein 1
Clonality Monoklonaal
Conjugatie Biotine
Clone Clone RBP/872
Reactivity Rabbit,Monkey,Goat,Human,Rat,Mouse
Host Mouse
Gen-ID 529571
Isotype IgG1 kappa
ImmunoChemie Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR