Anti-ALPP Mouse Monoclonal Antibody (CF660R) [clone: GM022]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC610874-500 BNC610874-100
BNC610874-500EA 560 EUR
BNC610874-500 BNC610874-100
Anti-ALPP Mouse Monoclonal Antibody (CF660R) [clone: GM022]
Antilichamen
Reacts with a 70 kDa membrane-bound isozyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. It is highly specific for PLAP and shows no cross-reaction with other isozymes of alkaline phosphatase. Anti-PLAP reacts with germ cell tumors and can discriminate between these and other neoplasms. Somatic neoplasms e.g. breast, gastrointestinal, prostatic, and urinary cancers may also immunoreact with antibodies to PLAP. Anti-PLAP positivity in conjunction with anti-keratin negativity favors seminoma over carcinoma.More Product Information