Anti-PTH Mouse Monoclonal Antibody (Purified) [clone: PTH/911]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNUB0911-500 BNUB0911-100
BNUB0911-500EA 511 EUR
BNUB0911-500 BNUB0911-100
Anti-PTH Mouse Monoclonal Antibody (Purified) [clone: PTH/911]
Antilichamen
The epitope of this MAb maps in the N-terminus of PTH, a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2 levels by dissolving the salts in bone and preventing their renal excretion.It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH);More Product Information