Anti-NGFR Mouse Monoclonal Antibody (CF568) [clone: NTR/912]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC680912-500 BNC680912-100
BNC680912-500EA 560 EUR
BNC680912-500 BNC680912-100
Anti-NGFR Mouse Monoclonal Antibody (CF568) [clone: NTR/912]
Antilichamen
This antibody recognizes a glycoprotein of 75 kDa, identified as low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). NGFR is expressed in various neural crest cells and their tumors such as melanocytes, melanomas, neuroblastomas, pheochromocytomas and neurofibromas. Reportedly, anti-NGFR is a reliable marker for desmoplastic and neurotropic melanomas. NGFR is expressed in mature non-neural cells such as perivascular cells, dental pulp cells, lymphoidal follicular dendritic cells, basal epithelium of oral mucosa and hair follicles, prostate basal cells, and myoepithelial cells. Anti-NGFR stains the myoepithelial cells of breast ducts and intra-lobular fibroblasts of breast ducts.

Type: Primary
Antigen: NGFR
Clonality: Monoclonal
Clone: NTR/912
Conjugation: CF568
Epitope:
Host: Mouse
Reactivity: Human, Non-human primates
Isotype: IgG1, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR