Anti-NGFR Mouse Monoclonal Antibody (CF640R) [clone: NTR/912]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC400912-500 BNC400912-100
BNC400912-500EA 560 EUR
BNC400912-500 BNC400912-100
Anti-NGFR Mouse Monoclonal Antibody (CF640R) [clone: NTR/912]
Antilichamen
This antibody recognizes a glycoprotein of 75 kDa, identified as low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). NGFR is expressed in various neural crest cells and their tumors such as melanocytes, melanomas, neuroblastomas, pheochromocytomas and neurofibromas. Reportedly, anti-NGFR is a reliable marker for desmoplastic and neurotropic melanomas. NGFR is expressed in mature non-neural cells such as perivascular cells, dental pulp cells, lymphoidal follicular dendritic cells, basal epithelium of oral mucosa and hair follicles, prostate basal cells,More Product Information
Order Now

Specificaties

antigeen symbool NGFR
Type antilichaam Nerve Growth Factor Receptor,Primair
Environmentally Preferable
Type antilichaam Nerve Growth Factor Receptor,Primair
Clonality Monoklonaal
Conjugatie CF® 640R
Clone Clone NTR/912
Reactivity Primates,Human
Host Mouse
Gen-ID 415768 , 681726
Isotype IgG1 kappa
ImmunoChemie Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR