Anti-NGFR Mouse Monoclonal Antibody (Biotin) [clone: NGFR5 NTR/912]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNCB0914-500 BNCB0914-100
BNCB0914-500EA 560 EUR
BNCB0914-500 BNCB0914-100
Anti-NGFR Mouse Monoclonal Antibody (Biotin) [clone: NGFR5 NTR/912]
Antilichamen
This antibody recognizes a glycoprotein of 75 kDa, identified as low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). NGFR is expressed in various neural crest cells and their tumors such as melanocytes, melanomas, neuroblastomas, pheochromocytomas and neurofibromas. Reportedly, anti-NGFR is a reliable marker for desmoplastic and neurotropic melanomas. NGFR is expressed in mature non-neural cells such as perivascular cells, dental pulp cells, lymphoidal follicular dendritic cells, basal epithelium of oral mucosa and hair follicles, prostate basal cells,More Product Information
Order Now

Specificaties

antigeen symbool NGFR
Type antilichaam Nerve Growth Factor Receptor,Primair
Environmentally Preferable
Type antilichaam Nerve Growth Factor Receptor,Primair
Clonality Monoklonaal
Conjugatie Biotine
Clone Clone NGFR5 NTR/912
Reactivity Primates,Human
Host Mouse
Gen-ID 415768 , 681726
Isotype IgG
ImmunoChemie Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR