Anti-HSP90B1 Mouse Monoclonal Antibody (CF405S) [clone: 9G10.F8.2]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC040940-500 BNC040940-100
BTIUBNC040940-500EA 560 EUR
BTIUBNC040940-500 BTIUBNC040940-100
Anti-HSP90B1 Mouse Monoclonal Antibody (CF405S) [clone: 9G10.F8.2]
Antilichamen
Recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.
Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: 9G10.F8.2
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Chicken, Cow, Dog, Guinea Pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis