Anti-MYADM Mouse Monoclonal Antibody (Biotin) [clone: MYADM/972]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNCB0972-500 BNCB0972-100
BNCB0972-500EA 560 EUR
BNCB0972-500 BNCB0972-100
Anti-MYADM Mouse Monoclonal Antibody (Biotin) [clone: MYADM/972]
Antilichamen
Recognizes a myeloid associated differentiation antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. It reacts with early precursor and mature forms of human and monkey myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.
Type: Primary
Antigen: MYADM
Clonality: Monoclonal
Clone: MYADM/972
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Macaque