Anti-SOX10 Mouse Monoclonal Antibody (CF647) [clone: SOX10/992]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC470992-500 BNC470992-100
BNC470992-500EA 560 EUR
BNC470992-500 BNC470992-100
Anti-SOX10 Mouse Monoclonal Antibody (CF647) [clone: SOX10/992]
Antilichamen
Recognizes a protein of ~55 kDa, identified as SOX10. This MAb is highly specific and does not cross-react with other members of the SOX-family. SOX genes comprise a family of genes that are related to the mammalian sex-determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. SOX-10 is a sensitive marker of melanoma, including conventional, spindled, and desmoplastic subtypes. It is expressed by metastatic melanomas and nodal capsular nevus in sentinel lymph nodes, but not by otherMore Product Information