Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB110-94162 NB110-94162SS
NOVUNB110-94162EA 591 EUR
NOVUNB110-94162 NOVUNB110-94162SS
Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody
Antilichamen
Rabbit Polyclonal MAT1/2A Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Rat, Bovine, Primate, Zebrafish, Mouse (-).

Type: Primary
Antigen: MNAT1/MAT2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR