Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (Purified) [clone: 246-B8.E7]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNUB1054-500 BNUB1054-100
BNUB1054-500EA 511 EUR
BNUB1054-500 BNUB1054-100
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (Purified) [clone: 246-B8.E7]
Antilichamen
Reacts with a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.
Type: Primary
Antigen: HLA-A; HLA-B; HLA-C
Clonality: Monoclonal
Clone: 246-B8.E7
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse, Non-human primates