Anti-PTPRC Mouse Monoclonal Antibody (CF640R) [clone: PTPRC/1147]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC401147-500 BNC401147-100
BNC401147-500EA 560 EUR
BNC401147-500 BNC401147-100
Anti-PTPRC Mouse Monoclonal Antibody (CF640R) [clone: PTPRC/1147]
Antilichamen
CD45R, also designated CD45 and PTPRC, has been identified as a transmembrane glycoprotein, broadly expressed among hematopoietic cells. Multiple isoforms of CD45R are distributed throughout the immune system according to cell type. These isoforms arise because of alternative splicing of exons 4, 5, and 6. The corresponding protein domains are characterized by the binding of monoclonal antibodies specific for CD45RA (exon 4), CD45RB (exon 5), CD45RC (exon 6) and CD45RO (exons 4 to 6 spliced out). The variation in these isoforms is localized to the extracellular domain of CD45R, while theMore Product Information