Anti-SUMO1 Mouse Monoclonal Antibody (Biotin) [clone: SUMO1/1188]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNCB1188-500 BNCB1188-100
BNCB1188-500EA 560 EUR
BNCB1188-500 BNCB1188-100
Anti-SUMO1 Mouse Monoclonal Antibody (Biotin) [clone: SUMO1/1188]
Antilichamen
This MAb is specific to SUMO-1 and shows no cross-reaction with either SUMO-2 or SUMO-3. The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, SUMO-2 and SUMO-3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesized as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2, and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteinsMore Product Information
Order Now

Specificaties

antigeen symbool Small ubiquitin-related modifier 1
Environmentally Preferable
Type antilichaam Primair,SUMO-1
Type antilichaam Primair,SUMO-1
Clonality Monoklonaal
Conjugatie Biotine
Clone Clone SUMO1/1188
Reactivity Human,Rat
Host Mouse
Gen-ID 596171
Isotype IgG1 kappa
Western blot Yes
ImmunoChemie Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR