Anti-KRT18 Mouse Monoclonal Antibody (Biotin) [clone: KRT18/1190]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNCB1190-500 BNCB1190-100
BNCB1190-500EA 560 EUR
BNCB1190-500 BNCB1190-100
Anti-KRT18 Mouse Monoclonal Antibody (Biotin) [clone: KRT18/1190]
Antilichamen
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It was reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine andMore Product Information
Order Now

Specificaties

antigeen symbool KRT18
Environmentally Preferable
Type antilichaam Primair,Cytokeratin 18
Antigeen synoniemen K18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 Protein
Type antilichaam Primair,Cytokeratin 18
Clonality Monoklonaal
Conjugatie Biotine
Clone Clone KRT18/1190
Reactivity Human,Rat
Host Mouse
Gen-ID 406013
Isotype IgG1 kappa
ImmunoChemie Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR