Anti-CELA3B Mouse Monoclonal Antibody (CF640R) [clone: CELA3B/1218]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC401218-500 BNC401218-100
BNC401218-500EA 560 EUR
BNC401218-500 BNC401218-100
Anti-CELA3B Mouse Monoclonal Antibody (CF640R) [clone: CELA3B/1218]
Antilichamen
This MAb recognizes a protein of ~17 kDa, identified as CELA3B (Chymotrypsin like elastase family member 3B). Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, elastase 3B has little elastolytic activity. Like most of the human elastases, elastase 3B is secreted from the pancreas as a zymogen and, like other serine proteases such as trypsin, chymotrypsin and kallikrein; it has a digestive function in theMore Product Information
Order Now

Specificaties

antigeen symbool CELA3B
Type antilichaam Primair,Chymotrypsin-like elastase family, member 3B
Environmentally Preferable
Antigeen synoniemen Elastase IIIB|ELA3B|Protease E|Chymotrypsin like elastase family member 3B (CELA3B)
Type antilichaam Primair,Chymotrypsin-like elastase family, member 3B
Clonality Monoklonaal
Conjugatie CF® 640R
Clone Clone CELA3B/1218
Reactivity Human
Host Mouse
Gen-ID 728752
Isotype IgG1 kappa
ELISA Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR