Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF405S) [clone: B22.1 B23.1]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC041221-100 BNC041221-500
BNC041221-100EA 203 EUR
BNC041221-100 BNC041221-500
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF405S) [clone: B22.1 B23.1]
Antilichamen
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This MAb cocktail recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 18 in the normal epidermis so only Paget cells are stained.Immunohistochemical staining with this MAb is indistinguishable from that obtained with monoclonal antibody 5D3.

Type: Primary
Antigen: KRT8 & KRT18
Clonality: Monoclonal
Clone: B22.1 B23.1
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR