Capillary columns, MXT®-Volatiles (Siltek®-treated stainless steel)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70922 70924 70925 70926 70927 70925-271
REST70922EA 0 EUR
REST70922 REST70924 REST70925 REST70926 REST70927 REST70925-271
Capillary columns, MXT®-Volatiles (Siltek®-treated stainless steel)
Kolommen voor chromatografie
MXT®-Volatiles columns are designed specifically for the analyses of 34 volatile organic pollutants listed in U.S. EPA methods 601, 602, and 624. With these columns, all compounds can be quantified using mass spectrometer or a PID in tandem with an ELCD.
  • Proprietary Crossbond® diphenyl dimethyl polysiloxane phase
  • Application-specific columns for volatile organic pollutants
  • Provides low bleed and thermal stability up to 320 °C