Capillary columns, Rxi®-20 (fused Silica)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10323 10324 10338 10339 10353 10354 10355
REST10323EA 720 EUR
REST10323 REST10324 REST10338 REST10339 REST10353 REST10354 REST10355
Capillary columns, Rxi®-20 (fused Silica)
Kolommen voor chromatografie
Rtx®-20 columns are general purpose columns for volatile compounds, flavour compounds, alcoholic beverages.
  • Low to midpolarity phase; Crossbond® diphenyl dimethyl polysiloxane
  • Temperature stability up to 320 °C
  • Equivalent to USP G28, G32 phases
Rtx®-20 polymer is synthesised to exacting standards. All residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight monomodal distribution and low bleed.