Molecular sieve columns (packed)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
80428-800 80428-810 80428-840 80429-800 80429-810 80429-820 80429-840 80430-800 80430-810 80430-840 80439-800 80439-810 80440-800 80440-810 80473-800 80473-810 80474-800 80474-810 80474-840 80475-800 80475-810 80475-820 80476-800 80476-820 80480-800 88014-800 88014-810 88015-800 88015-810 80439-820 80476-810
REST80428-800EA 531 EUR
REST80428-800 REST80428-810 REST80428-840 REST80429-800 REST80429-810 REST80429-820 REST80429-840 REST80430-800 REST80430-810 REST80430-840 REST80439-800 REST80439-810 REST80440-800 REST80440-810 REST80473-800 REST80473-810 REST80474-800 REST80474-810 REST80474-840 REST80475-800 REST80475-810 REST80475-820 REST80476-800 REST80476-820 REST80480-800 REST88014-800 REST88014-810 REST88015-800 REST88015-810 REST80439-820 REST80476-810
Molecular sieve columns (packed)
Kolommen voor chromatografie
Molecular sieve packed columns easily separate permanent gases at above-ambient temperatures.
  • Siltek®-treated stainless steel
  • Preactivated and ready to use
  • Excellent batch-to-batch reproducibility
Process of preparing molecular sieve packings results in excellent batch-to-batch reproducibility. In addition, these molecular sieves are preactivated and ready to use. Each column comes with metal end-fittings to prevent water or carbon dioxide from adsorbing into the packing during shipment.