Autoseal tip assembly for solvent extractor, ASE® 200/300 systems
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26161 26162 26246 26247
REST26161EA 160 EUR
REST26161 REST26162 REST26246 REST26247
Autoseal tip assembly for solvent extractor, ASE® 200/300 systems
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These assemblies meet original equipment manufacturer's performance.
Choose original equipment equivalent stainless steel or Siltek deactivation for improved inertness and extended lifetime.