Extraction cell caps and replacement parts
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26100 26187 26134 25959 25997 26096 26166 26174 26180 26184 26188 26169
REST26100EA 74 EUR
REST26100 REST26187 REST26134 REST25959 REST25997 REST26096 REST26166 REST26174 REST26180 REST26184 REST26188 REST26169
Extraction cell caps and replacement parts
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Caps features a smooth inner surfaces for easier cleaning. Suitable for for ASE systems.
Leveringsinformatie: Caps include frit, PEEK washer, PTFE O-ring, threaded insert and snap ring.