Extraction cell bodies for solvent extraction, ASE® systems
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26176 26178 25993 25994 25995 25996 26098 26112 26114 26116 26132
REST26176EA 152.5 EUR
REST26176 REST26178 REST25993 REST25994 REST25995 REST25996 REST26098 REST26112 REST26114 REST26116 REST26132
Extraction cell bodies for solvent extraction, ASE® systems
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Accelerated solvent extraction is a common technique for fast and reliable extraction of organic materials from solid matrices, using EPA SW-846 method 3545 and pressurised fluid extraction (PFE).
  • Cell bodies are serialised for easy sample identification
  • Smooth inner surfaces for easier cleaning
Bestelinformatie: Caps are sold separately.