Precision sampling valves for vials, Mininert®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24900 24901 24902 24903 24904 24905 24906 24907
REST24900EA 209 EUR
REST24900 REST24901 REST24902 REST24903 REST24904 REST24905 REST24906 REST24907
Precision sampling valves for vials, Mininert®
Ventielen Terugslagventielen
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
  • Available for screw cap and crimp top vials
  • Threaded flange used to secure tight fit