Poly crimp seal caps, 11 mm
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21728 24433 24434 24443 24444 24445 24446 24453 24454 24455 24456
REST21728EA 182 EUR
REST21728 REST24433 REST24434 REST24443 REST24444 REST24445 REST24446 REST24453 REST24454 REST24455 REST24456
Poly crimp seal caps, 11 mm
Sluitingen Sluitingen voor chromatografievials
The Poly Crimp seal is versatile, working as either a snap-on or crimp-top cap. The standard crimping tool can be used to secure the Poly Crimp seal onto any crimp-top vial.