Lockout station, Industrial Strength
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
800123 800129
BRDY800123EA 289.73 EUR
BRDY800123 BRDY800129
Lockout station, Industrial Strength
Veiligheidssloten
This lockable cabinet comes with one adjustable shelf (extra shelves are available separately).
  • Made of durable, powder-coated metal
  • Can be wall mounted or easily transported
  • Lockout Safety tips are printed on inside of the door
  • Components not included