All-Weather Vinyl Labels B-595 for BBP™31 Printers, Pre-Printed or Blank Headers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
142196 142198 142199 142188 142189 142190 142194 142195 142197 142182 142183 142184 142185 142186 142187 142193
BRDY142196EA 190.3 EUR
BRDY142196 BRDY142198 BRDY142199 BRDY142188 BRDY142189 BRDY142190 BRDY142194 BRDY142195 BRDY142197 BRDY142182 BRDY142183 BRDY142184 BRDY142185 BRDY142186 BRDY142187 BRDY142193
All-Weather Vinyl Labels B-595 for BBP™31 Printers, Pre-Printed or Blank Headers
Etiketten
Pre-printed pre-cut blank sign headers allow the printer to create multicolor signs fast - in a single print pass.
  • Durable, low-shrink vinyl with most aggressive adhesive
  • Conforms to irregular, curved, rough and highly textured surfaces
  • Adheres to 'difficult' low-surface energy items (PVC piping, blow-molded equipment cases, ABS plastics, recycled plastics)
Certificaten: Headers meet OSHA 1910.145 or ASME/ANSI Z535 standards.