Labels for BBP3x printers, type B-549
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
117865 117860 117861 117862 117863 117864
BRDY117865EA 211.43 EUR
BRDY117865 BRDY117860 BRDY117861 BRDY117862 BRDY117863 BRDY117864
Labels for BBP3x printers, type B-549
Etiketten
Cold temperature tape permanently adheres to surfaces at temperatures as low as −10 °F.
  • Ideal for use in any cold storage warehouse, cooler or freezer
  • Not recommended for plotter cutting