CYTAG™ CGH labelling kits
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZ42671K010 ENZ42671K100
ENZOENZ42671K010EA 873 EUR
ENZOENZ42671K010 ENZOENZ42671K100
CYTAG™ CGH labelling kits
Reagentia voor elektroforese Western Blotting Reagents
The CYTAG™ CGH labeling kit produces high quality data for comparative genomic hybridisation (aCGH) using as little as 0,25 µg of genomic DNA, without a need for pre-amplification.
  • Ability to perform total genomic DNA analysis without amplification or complexity reduction
  • Provides excellent dynamic analytical range for challenging and complex, heterogeneous samples
  • QC benchmarked and validated using high resolution array
  • Superior labeling efficiency and better dye incorporation results in less failed runs
Comparative genomic hybridisation (aCGH) is a powerful diagnostic tool for detecting DNA copy number gains and losses associated with chromosome abnormalities. aCGH provides an understanding of genetic disorders, cancers and other genomic aberrations.
Leveringsinformatie: Kits contain primers/reaction buffer, 1×400 µl or 1×4 ml; cyanine 3-dUTP nucleotide mix, 2×50 µl or 1×1 ml; cyanine 5-dUTP nucleotide mix, 2×50 µl or 1×1 ml; klenow exo-DNA polymerase, 1×20 µl or 1×200 µl; stop buffer, 1×100 µl or 1×1 ml; and nuclease-free water, 1×1 ml or 1×10 ml.