Wafer tweezers, without teeth
Leverancier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2LB/4 2LB-P
OUTI2LB/4EA 36.6 EUR
OUTI2LB/4 OUTI2LB-P
Wafer tweezers, without teeth
Pincetten
Stainless steel. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
  • Stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping
  • Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
  • Satin finish on the outside of the paddel for glare prevention
  • Each Wafer size has its corresponding tweezers size (e.g. for an 8" Wafer take tweezers with a No. 8)