Septum packed purge port weldment, Siltek®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21685 21691
REST21685EA 24 EUR
REST21685 REST21691
Septum packed purge port weldment, Siltek®
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
This Siltek® treated septum packed purge port weldment for Agilent 5890 GC makes weldment inert and eliminates adsorption of sensitive compounds such as, DDT and Endrin.
Bestelinformatie: Viton O-rings and appropriate septa have to be ordered separately.