FID collector assembly kit
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21001 21131 21132 21135 21136 21137 21138 21139 21699 23010
REST21001EA 84 EUR
REST21001 REST21131 REST21132 REST21135 REST21136 REST21137 REST21138 REST21139 REST21699 REST23010
FID collector assembly kit
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These FID collector assembly kits are made of high-quality stainless steel. Ideal for Agilent 5890/6850/6890/7890 GCs.