WorkHorse™ Labels for Thermal Transfer Printers B-488
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
217080 217091 217079 217085 062372 019261 062373 116381 029896 029893 029897 029895 116383
BRDY217080EA 272.1 EUR
BRDY217080 BRDY217091 BRDY217079 BRDY217085 BRDY062372 BRDY019261 BRDY062373 BRDY116381 BRDY029896 BRDY029893 BRDY029897 BRDY029895 BRDY116383
WorkHorse™ Labels for Thermal Transfer Printers B-488
Etiketten
Printable, high-performance polyester labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and AGA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.