Gas-tight syringes, PTFE Tip (Hamilton)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24558 24560 24562 24567 24568 24578 24591 24724 20178 20179
REST24558EA 129 EUR
REST24558 REST24560 REST24562 REST24567 REST24568 REST24578 REST24591 REST24724 REST20178 REST20179
Gas-tight syringes, PTFE Tip (Hamilton)
Spuiten Injectiespuiten voor chromatografie
These gas-tight syringes with interchangeable barrels and plungers are suitable for gases or liquids.
  • High accuracy of dispensed volumes
  • Tips with extend performance
  • Increase cost effectiveness
Bestelinformatie: Needles have to be ordered separately, for syringes without needles.