Cleaning rags in bags, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WIPE552 WIPE553
NEWPWIPE552EA 16.5 EUR
NEWPWIPE552 NEWPWIPE553
Cleaning rags in bags, PIG®
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
These recycled, white tee shirts work great for any general-purpose task, especially with solvents, which could cause coloured rags to bleed.
  • Made out of recycled, post-consumer, cotton-rich T-shirts and hosiery
  • Highly absorbent; works will with oil, ink and paint
  • Environmentally conscious products give old textiles a new life as strong and absorbent Rags, limiting the amount of textiles entering landfills
  • All rags are free of printing, metal content and areas of the garment that are not suitable for use such as cuffs and collars
  • Poly bag means there's no
More Product Information
Order Now
Specificaties
Environmentally Preferable