Laboratory step ladders, aluminium
Leverancier: GUENZBURGER STEIGTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11224 11225 11226 11227 11223
139-0470EA 0 EUR
139-0470 139-0471 139-0472 139-0473 139-0469
Laboratory step ladders, aluminium
Ladders en opstapjes
Step ladders made from aluminium. 15 years of quality guaranteed.
  • Slim-line construction for space saving storage
  • High stability due to conical design
  • Payload up to 150 kg
  • Corrugated aluminium steps (80 mm deep), ensuring a safe stance
  • Large, corrugated platform for safe working
  • Non slip corrugated ladder feet to ensure full contact with the underlying surface
  • Extra light and easy to use
Order Now
Specificaties
Environmentally Preferable