Packed column FID replacement jets
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21686 21687 21688 21689 21694 21695 21696 21697
REST21686EA 106 EUR
REST21686 REST21687 REST21688 REST21689 REST21694 REST21695 REST21696 REST21697
Packed column FID replacement jets
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These capillary dedicated FID replacement jet for Agilent 5890/6850/6890 GCs are available in standard, high-temperature or Siltek treated high-performance versions.
  • 0,46 mm ID jets are used for most general-purpose packed column applications
  • 0,76 mm ID jets are used for packings that exhibit high bleed and that frequently clog the tip of smaller 0,46 mm jets
The standard version threads are specially coated for easy installation and removal. Also special processing ensures the highest degree of cleanliness. High-performance version - Siltek treatedMore Product Information