Ionselectieve elektroden, natrium, Orion™ ROSS™ Sure-Flow™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8411BN 8611BNWP
662-0747EA 900 EUR
662-0747 662-1523
Ionselectieve elektroden, natrium, Orion™ ROSS™ Sure-Flow™
Elektroden ISE-elektroden
These electrodes are designed for measuring sodium in aqueous solutions quickly, accurately and economically.
  • Orion™ ROSS™ electrode provides faster response, greater stability and accuracy than conventional sodium electrodes
  • Combination electrode 8611BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes
  • Built-in Sure-Flow reference junction prevents electrode clogging and provides fast and stabile readings
Bestelinformatie: The half cell electrodes require a separate reference electrode which can be ordered separately.