Direct injection glass inserts
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20967 20967-200.5 20967-202.5 20967-210 20967-214
REST20967EA 39 EUR
REST20967 REST20967-200.5 REST20967-202.5 REST20967-210 REST20967-214
Direct injection glass inserts
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
These direct injection glass inserts are suitable for 6,35 mm packed column injection ports.
  • Closely controlled tolerance
  • Can be removed from the septum nut weldment