Purge and trap spargers
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21150 21151 26138 26139 26140 26141
REST21150EA 167 EUR
REST21150 REST21151 REST26138 REST26139 REST26140 REST26141
Purge and trap spargers
Filters Filters voor vloeistofverwerking
These purge and trap spargers are designed for use with Tekmar purge-and-trap instruments only and manufactured to tight tolerances to ensure a leak-tight seal.
  • Available with uniform frits to ensure maximum purging efficiency
  • Non-fritted spargers can be used for wastewater samples
  • With 12,7 mm mount