Vu-tight direct injection liners
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20343 20344 20787 20788 20342-202.1 20342-210.1 20342-214.1 20343-200.5 20343-202.5 20343-209.5 20343-213.5 20343-214.5 20344-209.25 20787-214.1 20788-214.5 20342
REST20343EA 282 EUR
REST20343 REST20344 REST20787 REST20788 REST20342-202.1 REST20342-210.1 REST20342-214.1 REST20343-200.5 REST20343-202.5 REST20343-209.5 REST20343-213.5 REST20343-214.5 REST20344-209.25 REST20787-214.1 REST20788-214.5 REST20342
Vu-tight direct injection liners
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
These Vu-tight Direct injection (DI) liners are suitable for 6,35 mm packed injection port conversion. They are available in two designs and fit 0,32 and 0,53 mm ID capillary columns (column outer diameter from 0,5 mm to 0,8 mm).
  • Visually observe the press-tight connection between the column end and liner
  • Slotted top prevents obstruction of carrier gas flow
  • Operate in the direct injection mode
Vu-tight DI liners can easily be packed with wool for dirty samples and cyclo Vu-Tight DI liners are ideal for dirty samples and spiral bore prevents non-volatile residue from contaminating the column.