Spill kit trolley bags, PIG® Oil-Only
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KITE250 KITE450
121-0506EA 204 EUR
121-0506 121-0569
Spill kit trolley bags, PIG® Oil-Only
Absorptiematerialen en bescherming tegen morsen
The spill kit trolley bag is intended for contingency planning and quick response to oil spills - but not water.
  • Luminous yellow bag is highly visible
  • Durable vinyl bag with hook-and-loop closure keeps absorbents dry and allows easy access
  • Wheeled trolley allows the kit to be easily manouvered to the spill site
Spill kits should be strategically placed at different locations in the workplace where spills are likely to happen.
Leveringsinformatie: 40× medium weight universal mat pads (380×510 mm), 6× socks (80×1220 mm), 8× pillows (250×250 mm), 5×More Product Information