Dubbelspatel, Chattaway, roestvrij staal
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2310 2311 2313 2315 2312 2314 2302 2303 2305 2317 2318
231-0078EA 11 EUR
231-0078 231-0360 231-0362 231-0363 231-0361 231-0342 231-0340 231-0341 NICK2305 231-0343 231-0344
Dubbelspatel, Chattaway, roestvrij staal
Spatels
Multipurpose spatula made of stainless steel for a wide range of uses. One end is flat and spatula-shaped, the other end is rounded and turned up at 45°. Available in light or medium weight design.
  • Very good chemical resistance
  • Very good thermal resistance
  • Quick and easy to clean