Graphite sealing rings
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21898 21899
REST21898EA 37 EUR
REST21898 REST21899
Graphite sealing rings
Afdichtingen Mechanische afdichtingen
These graphite sealing rings are suitable for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL instruments. They fit liners with 8 mm outer diameter.