Capillary columns, FAMEWAX (fused silica)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12497 12498 12499
REST12497EA 749 EUR
REST12497 REST12498 REST12499
Capillary columns, FAMEWAX (fused silica)
Kolommen voor chromatografie
Polar phase, Crossbond® polyethylene glycol application-specific columns for FAMEs, specially tested with a FAME mixture.
  • Temperature ranges from 20 to 250 °C
  • Comparably baseline resolution is achieved in significantly less time