Capillary columns, MXT®-WAX
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70620 70622 70623 70624 70625 70637 70638 70639 70640 70642 70651 70652 70654 70655 70657 70658 70666 70667 70669 70670 70672 70625-273 70640-271 70652-271 70655-271 70655-273
REST70620EA 558 EUR
REST70620 REST70622 REST70623 REST70624 REST70625 REST70637 REST70638 REST70639 REST70640 REST70642 REST70651 REST70652 REST70654 REST70655 REST70657 REST70658 REST70666 REST70667 REST70669 REST70670 REST70672 REST70625-273 REST70640-271 REST70652-271 REST70655-271 REST70655-273
Capillary columns, MXT®-WAX
Kolommen voor chromatografie
Polar phase general-purpose columns, with Crossbond® Carbowax® polyethylene glycol provides oxidation resistance.
  • Siltek®-treated stainless steel used
  • Temperature ranges from 40 to 260 °C
General-purpose columns for FAMEs, flavor compounds, essential oils, amines, solvents, xylene isomers, and U.S. EPA Method 603 (acrolein/acrylonitrile)