Solid black and white polystyrene microplates, 384-well, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3676 3570 3571 3572 3573 3574 3574BC 3575 3575BC 3576 3577 3820 3821 3821BC 3822 3824 3824BC 3825 3826 3826BC 3570BC 4511 4512 4513 4514
732-5533EA 910.47 EUR
732-5533 734-4104 734-4105 734-4106 734-4107 734-4108 734-4109 734-4110 734-4111 734-4112 734-4113 734-4142 734-4143 734-4144 734-4145 734-4146 734-4147 734-4148 734-4149 734-4150 738-0148 CORN4511 738-0213 738-0214 738-0215
Solid black and white polystyrene microplates, 384-well, Corning®
Microplaten HTS-microplaten
PS. Ontworpen om interferentie tussen wells te verminderen bij fluorescentie- en luminescentieanalysen.
  • Square well top for easier robotic pipetting
  • Round bottom for better Z factor and minimized trapped air
  • Conical well molded in the shape of a light cone for efficiency
  • Black walls reduce well-to-well crosstalk and autofluorescence during fluorescent assays
High binding surface is a general purpose surface that binds medium (>10kD) and large biomolecules possessing ionic groups and/or hydrophobic regions. Binding capacity is approximately 400 to 50More Product Information