Capillary columns, Rtx®-BAC2 (fused silica)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18000 18002 18000-6850 18002-6850
REST18000EA 821 EUR
REST18000 REST18002 REST18000-6850 REST18002-6850
Capillary columns, Rtx®-BAC2 (fused silica)
Kolommen voor chromatografie
Application-specific columns for blood alcohol analysis - achieve baseline resolution in less than three minutes.
  • Ideal for abused inhalant anesthetics, γ-hydroxybutyrate (GHB)/γ-butyrolactone (GBL), glycols, and common industrial solvents
  • Rtx®-BAC2 confirmation column provides four elution order changes under the same conditions
  • Stable to 260 °C